Assay Details
Gene Symbol: FABP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489404.4 | XP_001489454.1 | 311 | 96 | ||
XM_023648390.1 | XP_023504158.1 | 393 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
