Assay Details
Gene Symbol: PKIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003365598.4 | XP_003365646.1 | 301 | 85 | ||
XM_005613045.3 | XP_005613102.1 | 447 | 85 | |||
XM_023648432.1 | XP_023504200.1 | 190 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
