Assay Details
Gene Symbol: LOC100052030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648506.1 | XP_023504274.1 | 1148 | 103 | ||
XM_023648507.1 | XP_023504275.1 | 1147 | 103 | |||
XM_023648508.1 | XP_023504276.1 | 1346 | 103 | |||
XM_023648509.1 | XP_023504277.1 | 1160 | 103 | |||
XM_023648510.1 | XP_023504278.1 | 1109 | 103 |

More Information
Set Membership: |
![]() ![]() |
