Assay Details
Gene Symbol: LOC100052030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648506.1 | XP_023504274.1 | 1360 | 71 | ||
XM_023648507.1 | XP_023504275.1 | 1359 | 71 | |||
XM_023648508.1 | XP_023504276.1 | 1558 | 71 | |||
XM_023648509.1 | XP_023504277.1 | 1372 | 71 | |||
XM_023648510.1 | XP_023504278.1 | 1321 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
