Assay Details
Gene Symbol: LOC100052030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648508.1 | XP_023504276.1 | 331 | 117 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023648508.1 transcripts.
Set Membership: |
![]() ![]() |
