Assay Details
Gene Symbol: MTFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495100.4 | XP_001495150.1 | 1709 | 69 | ||
XM_023648531.1 | XP_023504299.1 | 1145 | 69 | |||
XM_023648532.1 | XP_023504300.1 | 1716 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
