Assay Details
Gene Symbol: GGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495840.5 | XP_001495890.1 | 455 | 78 | ||
XM_005613092.3 | XP_005613149.1 | 400 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
