Assay Details
Gene Symbol: RAB2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648555.1 | XP_023504323.1 | 723 | 98 | ||
GenBank mRNA | JL616161.2 | - | 5 - 6 | 380 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
