Assay Details
Gene Symbol: SDR16C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005613117.3 | XP_005613174.2 | 870 | 89 | ||
XM_005613119.3 | XP_005613176.2 | 697 | 89 | |||
XM_014728000.2 | XP_014583486.2 | 826 | 89 | |||
XM_023648572.1 | XP_023504340.1 | 826 | 89 | |||
XM_023648574.1 | XP_023504342.1 | 826 | 89 | |||
XM_023648575.1 | XP_023504343.1 | 694 | 89 | |||
XM_023648576.1 | XP_023504344.1 | 826 | 89 | |||
XM_023648577.1 | XP_023504345.1 | 826 | 89 | |||
XM_023648578.1 | XP_023504346.1 | 694 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
