Assay Details
Gene Symbol: MRPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648605.1 | XP_023504373.1 | 4452 | 79 | ||
XM_023648606.1 | XP_023504374.1 | 3302 | 79 | |||
XM_023648607.1 | XP_023504375.1 | 2432 | 79 | |||
XM_023648608.1 | XP_023504376.1 | 1962 | 79 | |||
XM_023648609.1 | XP_023504377.1 | 573 | 79 | |||
GenBank mRNA | JL622224.1 | - | 1 - 2 | 131 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
