Assay Details
Gene Symbol: RGS20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488849.4 | XP_001488899.1 | 724 | 69 | ||
XM_014728017.2 | XP_014583503.1 | 868 | 69 | |||
XM_014728018.2 | XP_014583504.1 | 769 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
