Assay Details
Gene Symbol: PCMTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005613177.3 | XP_005613234.1 | 832 | 122 | ||
XM_014728021.2 | XP_014583507.1 | 598 | 122 | |||
XM_023648627.1 | XP_023504395.1 | 876 | 122 | |||
XM_023648628.1 | XP_023504396.1 | 406 | 122 | |||
GenBank mRNA | JL623952.1 | - | 1 - 2 | 307 | 122 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014728021.2, XM_023648628.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
