Assay Details
Gene Symbol: TRIQK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005613194.3 | XP_005613251.1 | 306 | 71 | ||
XM_023648679.1 | XP_023504447.1 | 306 | 71 | |||
XM_023648681.1 | XP_023504449.1 | 306 | 71 | |||
XM_023648683.1 | XP_023504451.1 | 306 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005613194.3, XM_023648679.1, XM_023648681.1, XM_023648683.1 transcripts.
Set Membership: |
![]() ![]() |
