Assay Details
Gene Symbol: TRIQK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005613193.3 | XP_005613250.1 | 473 | 67 | ||
XM_005613194.3 | XP_005613251.1 | 707 | 67 | |||
XM_014728049.2 | XP_014583535.1 | 470 | 67 | |||
XM_023648679.1 | XP_023504447.1 | 821 | 67 | |||
XM_023648680.1 | XP_023504448.1 | 587 | 67 | |||
XM_023648681.1 | XP_023504449.1 | 885 | 67 | |||
XM_023648683.1 | XP_023504451.1 | 704 | 67 | |||
XM_023648684.1 | XP_023504452.1 | 537 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
