Assay Details
Gene Symbol: SDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915199.4 | XP_001915234.1 | 843 | 102 | ||
XM_023648735.1 | XP_023504503.1 | 981 | 102 | |||
XM_023648736.1 | XP_023504504.1 | 277 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
