Assay Details
Gene Symbol: POLR2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003365645.4 | XP_003365693.1 | 363 | 91 | ||
XM_005613229.3 | XP_005613286.1 | 422 | 91 | |||
XM_014728217.2 | XP_014583703.1 | 391 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
