Assay Details
Gene Symbol: ENY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495427.6 | XP_001495477.1 | 380 | 121 | ||
XM_003365620.4 | XP_003365668.1 | 375 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
