Assay Details
Gene Symbol: ZHX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648915.1 | XP_023504683.1 | 495 | 86 | ||
XM_023648916.1 | XP_023504684.1 | 1062 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023648915.1, XM_023648916.1 transcripts.
Set Membership: |
![]() ![]() |
