Assay Details
Gene Symbol: ZHX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648915.1 | XP_023504683.1 | 3345 | 99 | ||
XM_023648916.1 | XP_023504684.1 | 3912 | 99 | |||
XM_023648917.1 | XP_023504685.1 | 3393 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
