Assay Details
Gene Symbol: MYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497991.5 | XP_001498041.2 | 522 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001497991.5 transcripts.
Set Membership: |
![]() ![]() |
