Assay Details
Gene Symbol: CYRIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005613279.3 | XP_005613336.1 | 720 | 84 | ||
XM_023648972.1 | XP_023504740.1 | 917 | 84 | |||
XM_023648973.1 | XP_023504741.1 | 919 | 84 | |||
XM_023648974.1 | XP_023504742.1 | 633 | 84 | |||
GenBank mRNA | HQ890119.1 | - | 1 - 2 | 57 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
