Assay Details
Gene Symbol: PTP4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649030.1 | XP_023504798.1 | 1321 | 68 | ||
XM_023649031.1 | XP_023504799.1 | 1308 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
