Assay Details
Gene Symbol: GPIHBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496507.4 | XP_001496557.1 | 647 | 87 | ||
XM_005613335.3 | XP_005613392.1 | 431 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
