Assay Details
Gene Symbol: GLI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649110.1 | XP_023504878.1 | 553 | 73 | ||
XM_023649111.1 | XP_023504879.1 | 465 | 73 | |||
XM_023649112.1 | XP_023504880.1 | 559 | 73 |
