Assay Details
Gene Symbol: TBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648093.1 | XP_023503861.1 | 2757 | 63 | ||
XM_023648094.1 | XP_023503862.1 | 2148 | 63 | |||
XM_023648095.1 | XP_023503863.1 | 1053 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
