Assay Details
Gene Symbol: CCDC188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005613848.3 | XP_005613905.1 | 993 | 85 | ||
XM_014728647.2 | XP_014584133.1 | 993 | 85 | |||
XM_023646627.1 | XP_023502395.1 | 993 | 85 | |||
XM_023646628.1 | XP_023502396.1 | 993 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
