Assay Details
Gene Symbol: HECA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023633032.1 | XP_023488800.1 | 1567 | 73 | ||
XM_023633033.1 | XP_023488801.1 | 1473 | 73 | |||
XM_023633034.1 | XP_023488802.1 | 3375 | 73 | |||
GenBank mRNA | JL632919.1 | - | 1 - 2 | 68 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
