Assay Details
Gene Symbol: DHRSX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023633482.1 | XP_023489250.1 | 2112 | 55 | ||
XM_023633483.1 | XP_023489251.1 | 2121 | 55 | |||
XM_023633484.1 | XP_023489252.1 | 2022 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
