Assay Details
Gene Symbol: RPL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495765.5 | XP_001495815.3 | 363 | 86 | ||
XM_023650532.1 | XP_023506300.1 | 230 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
