Assay Details
Gene Symbol: CD99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023633870.1 | XP_023489638.1 | 290 | 54 | ||
XM_023633871.1 | XP_023489639.1 | 290 | 54 | |||
XM_023633872.1 | XP_023489640.1 | 366 | 54 | |||
XM_023633873.1 | XP_023489641.1 | 366 | 54 | |||
XM_023633874.1 | XP_023489642.1 | 290 | 54 | |||
XM_023633875.1 | XP_023489643.1 | 366 | 54 | |||
XM_023633876.1 | XP_023489644.1 | 290 | 54 | |||
XM_023633877.1 | XP_023489645.1 | 290 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
