Assay Details
Gene Symbol: AOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494989.5 | XP_001495039.3 | 2933 | 62 | ||
XM_014739402.2 | XP_014594888.2 | 2628 | 62 | |||
XM_023639979.1 | XP_023495747.1 | 3237 | 62 | |||
XM_023639980.1 | XP_023495748.1 | 2452 | 62 | |||
XM_023639981.1 | XP_023495749.1 | 2698 | 62 | |||
XM_023639982.1 | XP_023495750.1 | 2566 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
