Assay Details
Gene Symbol: ATG9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014739378.2 | XP_014594864.1 | 2161 | 66 | ||
XM_023639998.1 | XP_023495766.1 | 3367 | 66 | |||
XM_023639999.1 | XP_023495767.1 | 3248 | 66 | |||
XM_023640000.1 | XP_023495768.1 | 3293 | 66 | |||
XM_023640001.1 | XP_023495769.1 | 3217 | 66 | |||
XM_023640002.1 | XP_023495770.1 | 1880 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
