Assay Details
Gene Symbol: IQCA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005609556.3 | XP_005609613.1 | 75 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005609556.3 transcripts.
Set Membership: |
![]() ![]() |
