Assay Details
Gene Symbol: WDR86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640020.1 | XP_023495788.1 | 680 | 98 | ||
XM_023640021.1 | XP_023495789.1 | 674 | 98 | |||
XM_023640022.1 | XP_023495790.1 | 674 | 98 | |||
XM_023640023.1 | XP_023495791.1 | 675 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
