Assay Details
Gene Symbol: ETNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640325.1 | XP_023496093.1 | 1365 | 69 | ||
XM_023640326.1 | XP_023496094.1 | 1407 | 69 | |||
XM_023640327.1 | XP_023496095.1 | 1383 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
