Assay Details
Gene Symbol: MDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489035.4 | XP_001489085.3 | 1070 | 66 | ||
XM_023640360.1 | XP_023496128.1 | 1088 | 66 | |||
XM_023640362.1 | XP_023496130.1 | 1018 | 66 | |||
XM_023640363.1 | XP_023496131.1 | 1023 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
