Assay Details
Gene Symbol: RAB29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490736.6 | XP_001490786.1 | 227 | 85 | ||
XM_014739524.2 | XP_014595010.1 | 347 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
