Assay Details
Gene Symbol: RAB7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490878.4 | XP_001490928.1 | 299 | 60 | ||
XM_005609599.3 | XP_005609656.1 | 308 | 60 | |||
XM_023640427.1 | XP_023496195.1 | 320 | 60 | |||
XM_023640428.1 | XP_023496196.1 | 331 | 60 | |||
XM_023640429.1 | XP_023496197.1 | 304 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
