Assay Details
Gene Symbol: RAB7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490878.4 | XP_001490928.1 | 644 | 100 | ||
XM_005609599.3 | XP_005609656.1 | 653 | 100 | |||
XM_023640427.1 | XP_023496195.1 | 665 | 100 | |||
XM_023640429.1 | XP_023496197.1 | 649 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
