Assay Details
Gene Symbol: RASSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491727.4 | XP_001491777.1 | 609 | 72 | ||
XM_023640452.1 | XP_023496220.1 | 714 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
