Assay Details
Gene Symbol: ILDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640472.1 | XP_023496240.1 | 979 | 61 | ||
XM_023640473.1 | XP_023496241.1 | 1129 | 61 | |||
XM_023640474.1 | XP_023496242.1 | 977 | 61 | |||
XM_023640475.1 | XP_023496243.1 | 987 | 61 | |||
XM_023640476.1 | XP_023496244.1 | 986 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
