Assay Details
Gene Symbol: RCSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640487.1 | XP_023496255.1 | 345 | 56 | ||
XM_023640488.1 | XP_023496256.1 | 397 | 56 | |||
XM_023640489.1 | XP_023496257.1 | 255 | 56 | |||
XM_023640490.1 | XP_023496258.1 | 307 | 56 | |||
XM_023640491.1 | XP_023496259.1 | 3710 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
