Assay Details
Gene Symbol: MPZL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640492.1 | XP_023496260.1 | 618 | 68 | ||
XM_023640493.1 | XP_023496261.1 | 618 | 68 | |||
XM_023640494.1 | XP_023496262.1 | 618 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
