Assay Details
Gene Symbol: GPR161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005609634.3 | XP_005609691.1 | 2450 | 69 | ||
XM_023640506.1 | XP_023496274.1 | 1749 | 69 | |||
XM_023640507.1 | XP_023496275.1 | 1563 | 69 | |||
XM_023640508.1 | XP_023496276.1 | 2277 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
