Assay Details
Gene Symbol: VEGFD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489992.4 | XP_001490042.1 | 996 | 72 | ||
XM_023634142.1 | XP_023489910.1 | 1328 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
