Assay Details
Gene Symbol: MBTPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023634089.1 | XP_023489857.1 | 345 | 60 | ||
XM_023634090.1 | XP_023489858.1 | 344 | 60 | |||
XM_023634091.1 | XP_023489859.1 | 344 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Molecular Function -




