Assay Details
Gene Symbol: GK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003365769.3 | XP_003365817.2 | 1311 | 80 | ||
XM_014728796.2 | XP_014584282.1 | 1395 | 80 | |||
XM_014728797.2 | XP_014584283.1 | 1377 | 80 | |||
XM_014728798.2 | XP_014584284.1 | 1395 | 80 | |||
XM_014728799.2 | XP_014584285.1 | 1293 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
