Assay Details
Gene Symbol: TAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488600.6 | XP_001488650.1 | 2387 | 127 | ||
XM_005614055.3 | XP_005614112.1 | 2511 | 127 | |||
XM_005614057.3 | XP_005614114.1 | 2511 | 127 | |||
XM_005614058.3 | XP_005614115.1 | 2513 | 127 | |||
XM_023634094.1 | XP_023489862.1 | 2624 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
