Assay Details
Gene Symbol: ZNF41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005614095.3 | XP_005614152.1 | 148 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005614095.3 transcripts.
Set Membership: |
![]() ![]() |
