Assay Details
Gene Symbol: EBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493659.4 | XP_001493709.1 | 767 | 84 | ||
XM_005614110.3 | XP_005614167.1 | 683 | 84 | |||
XM_005614113.3 | XP_005614170.1 | 634 | 84 | |||
XM_014728835.2 | XP_014584321.1 | 512 | 84 | |||
XM_023633293.1 | XP_023489061.1 | 806 | 84 | |||
GenBank mRNA | HQ890182.1 | - | 4 - 5 | 544 | 84 | |
JL616032.1 | - | 4 - 5 | 645 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
